missing translation for 'onlineSavingsMsg'
Learn More

Geldanamycin, MedChemExpress

Product Code. 16466378
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16466378 10 mg 10mg
16458967 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16466378 Supplier Medchem Express Supplier No. HY15230
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Geldanamycin is a Hsp90 inhibitor with antimicrobial activity against many Gram-positive and some Gram-negative bacteria. Geldanamycin has anti-influenza virus H5N1 activities.

Geldanamycin is a Hsp90 inhibitor with antimicrobial activity against many Gram-positive and some Gram-negative bacteria. Geldanamycin has anti-influenza virus H5N1 activities.

Specifications

Chemical Name or Material Geldanamycin
Physical Form Solid
CAS 30562-34-6
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C29H40N2O9
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 50 mg/mL (89.18 mM; Need ultrasonic)
SMILES COC(C(C=C1NC(/C(C)=C/C=C/[C@H](OC)[C@@H](OC(N)=O)/C(C)=C/[C@H](C)[C@H]2O)=O)=O)=C(C[C@H](C[C@@H]2OC)C)C1=O
Molecular Weight (g/mol) 560.64
Formula Weight 560.64
Percent Purity 97.08%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.