missing translation for 'onlineSavingsMsg'
Learn More

Geldanamycin, 98+%

Product Code. 15425649
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15425649 250 mg 250mg
15405649 25 mg 25mg
15415649 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15425649 Supplier Thermo Scientific Alfa Aesar Supplier No. J63397.MD
 Restricted Item
Request Bulk or Custom Formats

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Benzoquinoid antibiotic that inhibits pp60src tyrosine kinase | CAS: 30562-34-6 | C29H40N2O9 | 560.644 g/mol

Geldanamycin binds to and inhibits the cytosolic chaperone functions of heat shock protein 90 (HSP90). HSP90 maintains the stability and functional shape of many oncogenic signaling proteins. It interferes with conformational maturation of proteins and the cellular stress response. In addition, it is a potent inhibitor of the nuclear hormone receptor family.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Geldanamycin binds to and inhibits the cytosolic chaperone functions of heat shock protein 90 (HSP90). HSP90 maintains the stability and functional shape of many oncogenic signaling proteins. It interferes with conformational maturation of proteins and the cellular stress response. In addition, it is a potent inhibitor of the nuclear hormone receptor family.

Solubility
Soluble in dimethyl sulfoxide at 100 mg/mL.

Notes
Store in a cool dry place. Keep away from oxidizing agents. Protect from heat.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 30562-34-6
Molecular Formula C29H40N2O9
Molecular Weight (g/mol) 560.644
MDL Number MFCD00274570
InChI Key QTQAWLPCGQOSGP-KDFVLELPSA-N
Synonym NSC 122750; U-29135
PubChem CID 133688416
IUPAC Name [(3S,5R,6S,7R,8Z,11R,14E)-6-hydroxy-5,11,21-trimethoxy-3,7,9,15-tetramethyl-16,20,22-trioxo-17-azabicyclo[16.3.1]docosa-1(21),8,12,14,18-pentaen-10-yl] carbamate
SMILES CC1CC(C(C(C=C(C(C(C=CC=C(C(=O)NC2=CC(=O)C(=C(C1)C2=O)OC)C)OC)OC(=O)N)C)C)O)OC

Specifikationer

Type Geldanamycin
Physical Form Powder
Color Orange to Yellow
Quantity 250 mg
Solubility Information Soluble in dimethyl sulfoxide at 100mg/mL.
Formula Weight 560.64
Percent Purity ≥99%
Chemical Name or Material Geldanamycin

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.