Learn More
Fisherbrand™ Supercooled Gel Tank, Complete - For Precast Or Handcast Gels
Click to view available options
Type:
Supercooled Gel Tank, Complete
Unit Size:
Each
Description
- A convenient and space saving solution for slab gel electrophoresis and electroblotting.
- Combined electrophoresis and blotting - a one-tank solution for consistency of slab gel electrophoresis and electroblotting
- Cooled vertical tank complete with casting base plus accessories, the electroblotting module and accessories
Specifications
Specifications
| Type | Supercooled Gel Tank, Complete |
| Includes | Supercooled gel tank, gel running module, 2 x plain glass plates with 1mm bondedspacers, 2 x notched glass plates, 2 x 1mm (24 sample) comb, 1 x dummy plate,casting base with seals and spacer aligners, internal blotting module, compressioncassettes and fibre pads (Maxi gel units 205mm x 200mm) |