missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GEFT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106412
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GEFT.
The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange. The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange (Souchet et al., 2002 ).
Specifications
| GEFT | |
| Polyclonal | |
| Unconjugated | |
| Arhgef25 | |
| 2410008H17Rik; ARHGEF25; D10Ertd610e; Geft; guanine nucleotide exchange factor GEFT; p63RhoGEF; RAC/CDC42 exchange factor; RAC/CDC42 exchange factor isoform 2; rac/Cdc42/Rho exchange factor GEFT; Rho guanine nucleotide exchange factor (GEF) 25; Rho guanine nucleotide exchange factor 25; RhoA/RAC/CDC42 exchange factor; rhoA/Rac/Cdc42 guanine nucleotide exchange factor GEFT | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 115557, 314904, 52666 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6P720, Q86VW2, Q9CWR0 | |
| Arhgef25 | |
| A synthesized peptide derived from human ARHGEF25(Accession Q86VW2), corresponding to amino acid residues V323-S373. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction