missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GEFT Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA562533

Product Code. 15700332

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: HSKHCLSVET EADSGQAGPY ENWMLEPALA TGEELPELTL LTTLLEGPGD KTQPPEEETL SQAPESEEEQ KKKALERSMY V Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange. The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange (Souchet et al., 2002 ).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GEFT
Polyclonal
Unconjugated
Arhgef25
2410008H17Rik; ARHGEF25; D10Ertd610e; Geft; guanine nucleotide exchange factor GEFT; p63RhoGEF; RAC/CDC42 exchange factor; RAC/CDC42 exchange factor isoform 2; rac/Cdc42/Rho exchange factor GEFT; Rho guanine nucleotide exchange factor (GEF) 25; Rho guanine nucleotide exchange factor 25; RhoA/RAC/CDC42 exchange factor; rhoA/Rac/Cdc42 guanine nucleotide exchange factor GEFT
Rabbit
Antigen Affinity Chromatography
RUO
115557
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q86VW2
Arhgef25
Recombinant protein corresponding to Human GEFT. Recombinant protein control fragment (Product #RP-100581).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.