missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GEFT Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-14309-25ul
This item is not returnable.
View return policy
Description
GEFT Polyclonal specifically detects GEFT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GEFT | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| GEFTRAC/CDC42 exchange factor, Guanine nucleotide exchange factor GEFT, p63RhoGEFrho guanine nucleotide exchange factor 25, Rac/Cdc42/Rho exchange factor GEFT, Rho guanine nucleotide exchange factor (GEF) 25, RhoA/RAC/CDC42 exchange factor, RhoA/Rac/Cdc42 guanine nucleotide exchange factor GEFT | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 115557 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ARHGEF25 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: HSKHCLSVETEADSGQAGPYENWMLEPALATGEELPELTLLTTLLEGPGDKTQPPEEETLSQAPESEEEQKKKALERSMYV | |
| 25ul | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction