missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Gefitinib

Product Code. 17130182
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17130182 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17130182 Supplier STEMCELL Technologies Supplier No. 73162
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tyrosine kinase inhibitor; Inhibits EGFR

  • Gefitinib is a selective inhibitor of epidermal growth factor receptor (EGFR) tyrosine kinase that binds competitively in the ATP binding pocket with IC50 values of 23 and 80 nM for A431 vulval squamous carcinoma cells and KB cells, respectively (Barker et al.)
  • Blocks proliferation in vitro and in mouse xenograft models, of multiple cancer cell types including colon, ovarian, and breast cancer cell lines (Ciardiello et al.)
  • Induces apoptosis in the HaCaT human keratinocyte cell line via a c-Jun N-terminal kinase (JNK) activation, an EGFR-independent mechanism (Lu et al.)

Specifications

CAS 184475-35-2
Synonym ZD1839; Gefonib
Quantity 500 mg
Molecular Formula C22H24ClFN4O3
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.