missing translation for 'onlineSavingsMsg'
Learn More

Gefitinib Morpholine N-oxide, TRC

Product Code. 30407285
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407285 50 mg 50mg
30393944 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30407285 Supplier Toronto Research Chemicals Supplier No. TRCG30400550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gefitinib N-Oxide
CAS 847949-51-3
Purity Grade Notes HPLC
Molecular Formula C22 H24 Cl F N4 O4
Quantity 50 mg
Synonym 4-Quinazolinamine, N-(3-chloro-4-fluorophenyl)-7-methoxy-6-[3-(4-oxido-4-morpholinyl)propoxy]-, Gefitinib N-Oxide, N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-[3-(4-oxido-4-morpholinyl)propoxy]-4-quinazolinamine, Gefitinib morpholine N-oxide
InChI Formula InChI=1S/C22H24ClFN4O4/c1-30-20-13-19-16(12-21(20)32-8-2-5-28(29)6-9-31-10-7-28)22(26-14-25-19)27-15-3-4-18(24)17(23)11-15/h3-4,11-14H,2,5-10H2,1H3,(H,25,26,27)
SMILES COc1cc2ncnc(Nc3ccc(F)c(Cl)c3)c2cc1OCCC[N+]4([O-])CCOCC4
IUPAC Name N-(3-chloro-4-fluorophenyl)-7-methoxy-6-[3-(4-oxidomorpholin-4-ium-4-yl)propoxy]quinazolin-4-amine
Molecular Weight (g/mol) 462.9
Formula Weight 462.147
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.