missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gefitinib hydrochloride, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
500mg
5g
Description
Gefitinib hydrochloride (ZD1839 hydrochloride) is a potent, selective and orally active EGFR tyrosine kinase inhibitor with an IC50 of 33 nM. Gefitinib hydrochloride selectively inhibits EGF-stimulated tumor cell growth (IC50 of 54 nM) and that blocks EGF-stimulated EGFR autophosphorylation in tumor cells. Gefitinib hydrochloride also induces autophagy. Gefitinib hydrochloride has antitumour activity.
Specifications
Specifications
| Chemical Name or Material | Gefitinib hydrochloride |
| CAS | 184475-55-6 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H25Cl2FN4O3 |
| Quantity | 100 mg |
| Synonym | ZD-1839 hydrochloride |
| Solubility Information | H2O : 6.25 mg/mL (12.93 mM; Need ultrasonic) ∣DMSO : 0.227 mg/mL (0.47 mM; Need ultrasonic and warming) |
| SMILES | ClC1=C(C=CC(NC2=NC=NC3=C2C=C(C(OC)=C3)OCCCN4CCOCC4)=C1)F.[H]Cl |
| Molecular Weight (g/mol) | 483.36 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction