missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gefitinib-based PROTAC 3, MedChemExpress
Description
Gefitinib-based PROTAC 3, conjugating an EGFR binding element to a von Hippel-Lindau ligand via a linker, induces EGFR degradation with DC50s of 11.7 nM and 22.3 nM in HCC827(exon 19 del) and H3255 (L858R mutantion) cells, respectively.
Specifications
Specifications
| Chemical Name or Material | Gefitinib-based PROTAC 3 |
| CAS | 2230821-27-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C47H57ClFN7O8S |
| Quantity | 5 mg |
| Solubility Information | DMSO : ≥ 60 mg/mL (64.20 mM) |
| SMILES | O=C([C@H]1N(C([C@@H](NC(CCOCCOCCCCCOC2=CC3=C(NC4=CC=C(F)C(Cl)=C4)N=CN=C3C=C2OC)=O)C(C)(C)C)=O)C[C@H](O)C1)NCC5=CC=C(C6=C(C)N=CS6)C=C5 |
| Molecular Weight (g/mol) | 934.51 |
| Formula Weight | 934.51 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?