missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GEF5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560645
This item is not returnable.
View return policy
Description
Immunogen sequence: QGEERKRELQ VPEENRADSQ DEKSQTFLGK SEEVTGKQED HGIKEKGVPV SGQEAKEPES WDGGRLGAVG RARSREEENE HHGPSMPALI APEDSPHCDL FP Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 30%.
This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.
Specifications
| GEF5 | |
| Polyclonal | |
| Unconjugated | |
| ARHGEF5 | |
| 2210412D05Rik; AA717842; ARH5; ARHGEF5; AW495314; DKFZp686N1969; Ephexin 3; Ephexin-3; GEF5; Guanine nucleotide regulatory protein TIM; oncogene TIM; P60; p60 TIM; Rho guanine nucleotide exchange factor (GEF) 5; Rho guanine nucleotide exchange factor 5; TIM; TIM1; Transforming immortalized mammary oncogene | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7984 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q12774 | |
| ARHGEF5 | |
| Recombinant protein corresponding to Human GEF5. Recombinant protein control fragment (Product #RP-104368). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction