missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GEF5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15684045
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15684045 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15684045 Supplier Invitrogen™ Supplier No. PA550117

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total ARHGEF5 protein.

This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GEF5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARHGEF5
Gene Accession No. E9Q7D5, Q12774
Gene Alias 2210412D05Rik; AA717842; ARH5; ARHGEF5; AW495314; DKFZp686N1969; Ephexin 3; Ephexin-3; GEF5; Guanine nucleotide regulatory protein TIM; oncogene TIM; P60; p60 TIM; Rho guanine nucleotide exchange factor (GEF) 5; Rho guanine nucleotide exchange factor 5; TIM; TIM1; Transforming immortalized mammary oncogene
Gene Symbols ARHGEF5
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human ARHGEF5.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54324, 7984
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.