missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GEF-H1 Recombinant Rabbit Monoclonal Antibody (23GB3675), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553443
This item is not returnable.
View return policy
Description
GEF-H1 activates Rho GTPases by promoting the exchange of GDP and GTP. Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. GEF-H1 may be involved in epithelial barrier permeability, cell motility and polarization, dendritic spine morphology, antigen presentation, leukemic cell differentiation, cell cycle regulation, innate immune response, and cancer. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| GEF-H1 | |
| Recombinant Monoclonal | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5FVC2, Q60875, Q92974 | |
| ARHGEF2 | |
| A synthetic peptide of human GEF H1 (950-986AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3675 | |
| Unconjugated | |
| ARHGEF2 | |
| AA408978; ARHGEF2; CG10188; CG10188-PA; CG10188-PB; CG10188-PC; cyst; cyst-PA; cyst-PB; cyst-PC; cysts; DKFZp547L106; DKFZp547P1516; Dmel\CG10188; Dmel_CG10188; FLJ45440; GEF; GEFH1; GEF-H1; Guanine nucleotide exchange factor H1; Kiaa0651; Lbcl1; LBC'S first cousin; Lfc; LFP40; Lymphoid blast crisis-like 1; microtubule-regulated Rho-GEF; mKIAA0651; oncogene LFC; OTTHUMP00000018855; P40; Proliferating cell nucleolar antigen p40; rho guanine nucleotide exchange factor 2; rho guanine nucleotide exchange factor 2-like protein; Rho/Rac guanine nucleotide exchange factor (GEF) 2; Rho/Rac guanine nucleotide exchange factor 2; rhobin | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16800, 310635, 9181 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction