missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GEF-H1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24472-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein containing a large tandem-repeat domain. The mouse ortholog of this gene has been shown to function in ciliogenesis in inner medullary collecting duct cells. Mutations in this gene have been associated with Alstrom syndrome. Alternative splice variants have been described but their full length sequences have not been determined.Specifications
| GEF-H1 | |
| Polyclonal | |
| Unconjugated | |
| ARHGEF2 | |
| ARHGEF2, GEF, GEF H1, GEFH1, GEF-H1, KIAA0651, LFP40, Microtubule regulated Rho GEF, P40 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9181 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q92974 | |
| ARHGEF2 | |
| GEF-H1 Fusion Protein Ag13352 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction