missing translation for 'onlineSavingsMsg'
Learn More

GEF-H1 Antibody, Novus Biologicals™

Product Code. 18389348 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389348 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389348 Supplier Novus Biologicals Supplier No. H00009181B01P50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

GEF-H1 Polyclonal antibody specifically detects GEF-H1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GEF-H1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. ABM85604.1
Gene Alias GEF, GEFH1, GEF-H1DKFZp547P1516, Guanine nucleotide exchange factor H1, KIAA0651DKFZp547L106, LFP40FLJ45440, Microtubule-regulated Rho-GEF, P40, Proliferating cell nucleolar antigen p40, rho guanine nucleotide exchange factor 2, Rho/Rac guanine nucleotide exchange factor (GEF) 2, Rho/Rac guanine nucleotide exchange factor 2
Host Species Mouse
Immunogen ARHGEF2 (ABM85604.1, 1 a.a. - 959 a.a.) full-length human protein. MKEAKDARYTNGHLFTTISVSGMTMCYACNKSITAKEALICPTCNVTIHNRCKDTLANCTKVKQKQQKAALLKNNTALQSVSLRSKTTIRERPSSAIYPSDSFRQSLLGSRRGRSSLSLAKSVSTTNIAGHFNDESPLGLRRILSQSTDSLNMRNRTLSVESLIDEAEVIYSELMSDFEMDEKDFAADSWSLAVDSSFLQQHKKEVMKQQDVIYELIQTELHHVRTLKIMTRLFRTGMLEELHLEPGVVQGLFPCVDELSDIHTRFLSQLLERRRQALCPGSTRNFVIHRLGDLLISQFSGPSAEQMCKTYSEFCSRHSKALKLYKELYARDKRFQQFIRKVTRPAVLKRHGVQECILLVTQRITKYPLLISRILQHSHGIEEERQDLTTALGLVKELLSNVDEGIYQLEKGARLQEIYNRMDPRAQTPVPGKGPFGREELLRRKLIHDGCLLWKTATGRFKDVLVLLMTDVLVFLQEKDQKYIFPTLDKPSVVSLQNLIVRDIANQEKGMFLISAAPPEMYEVHTASRDDRSTWIRVIQQSVRTCPSREDFPLIETEDEAYLRRIKMELQQKDRALVELLREKVGLFAEMTHFQAEEDGGSGMALPTLPRGLFRSESLESPRGERLLQDAIREVEGLKDLLVGPGVELLLTPREPALPLEPDSGGNTSPGVTANGEARTFNGSIELCRADSDSSQRDRNGNQLRSPQEEALQRLVNLYGLLHGLQAAVAQQDTLMEARFPEGPERREKLCRANSRDGEAGRAGAAPVAPEKQATELALLQRQHALLQEELRRCRRLGEERATEAGSLEARLRESEQARALLEREAEEARRQLAALGQTEPLPAEAPWARRPVDPRRRSLPAGDALYLSFNPPQPSRGTDRLDLPVTTRSVHRNFEDRERQELGSPEERLQDSSDPDTGSEEEGSSRLSPPHSPRDFTRMQDIPEETESRDGEAVASES
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9181
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.