Learn More
Gedivumab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Brand: Invitrogen MA542145
Additional Details : Weight : 0.01000kg
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Gedivumab is under investigation in clinical trial NCT01877785 (A Study of MHAA4549A to Assess Safety And Pharmacokinetics in Healthy Volunteers).Specifications
Gedivumab | |
Recombinant Monoclonal | |
Unconjugated | |
MHAA4549A; RO6876802;RG7745 | |
Influenza A virus hemagglutinin HA | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
1.4 mg/mL | |
PBS with no preservative, pH 7.5 | |
Human | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG1 κ |