missing translation for 'onlineSavingsMsg'
Learn More

GeA-69, MedChemExpress

Product Code. 16470567
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16470567

Brand: Medchem Express HY108708

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GeA-69 is a selective, allosteric inhibitor of poly-adenosine-diphosphate-ribose polymerase 14 (PARP14) targeting macrodomain 2, with a Kd of 2.1 μM.

GeA-69 is a selective, allosteric inhibitor of poly-adenosine-diphosphate-ribose polymerase 14 (PARP14) targeting macrodomain 2, with a Kd of 2.1 μM.

Specifications

Chemical Name or Material GeA-69
Physical Form Solid
CAS 2143475-98-1
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H16N2O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 125 mg/mL (416.18 mM)
SMILES CC(NC1=CC=CC=C1C2=CC=CC3=C2NC4=C3C=CC=C4)=O
Molecular Weight (g/mol) 300.35
Formula Weight 300.35
Percent Purity 99.97%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.