missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sepharose Agarose Gel Filtration Media

Product Code. 10359864 Shop All Cytiva Products
Change view
Click to view available options
Flow Rate:
<10 cm/hr.
<11.5 cm/hr.
<14 cm/hr.
<15 cm/hr.
<26 cm/hr.
<30 cm/hr.
Particle Size:
45 to 165 μm
60 to 200 μm
pH Range:
3 to 12
3 to 13
4 to 9
Support Type:
2% Agarose
4% Agarose
6% Agarose
Unit Size:
1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Particle Size pH Range Support Type unitSize
10359864 <10 cm/hr. 60 to 200 μm 4 to 9 2% Agarose 1 L
10004634 <26 cm/hr. 45 to 165 μm 3 to 13 2% Agarose 1 L
10217754 <15 cm/hr. 60 to 200 μm 3 to 13 2% Agarose 1 L
10605355 <11.5 cm/hr. 45 to 165 μm 4 to 9 4% Agarose 1 L
10702627 <30 cm/hr. 45 to 165 μm 3 to 12 6% Agarose 1 L
10695345 <14 cm/hr. 45 to 165 μm 4 to 9 6% Agarose 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10359864 Supplier Cytiva Supplier No. 17013001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Choose from a wide range of bead-formed agarose-based gel filtration media. Cytiva Life Sciences™ Sepharose Agarose Gel Filtration Media shows good separation over a wide molecular weight range.

European Pharmacopoeia Solutions

  • Reagents and Standard solutions in Chapters 2 and 4 of the current Ph. Eur.
  • All are manufactured and tested in compliance with the Ph. Eur.
  • Supplied with a Certificates of Analysis
  • Lot No. and expiry date are stated on all product labels

Order Info

Supplied in 1L plastic containers

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 L
Flow Rate <10 cm/hr.
Particle Size 60 to 200 μm
Product Type Sepharose Agarose Gel Filtration Media
Format Solid Bead
For Use With (Application) Allow characterization or cleaning up of samples containing components of diverse molecular weight
pH Range 4 to 9
Support Type 2% Agarose
Content And Storage 4° to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.