missing translation for 'onlineSavingsMsg'
Learn More

Cytiva rmp Protein A Sepharose™ Fast Flow Affinity Media

Product Code. 10358374 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
200 mL
Unit Size:
200mL
25mL
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10358374 200 mL 200mL
11369981 5 mL Each
11349981 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10358374 Supplier Cytiva Supplier No. 17513803

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed for high-purity separation of monoclonal and polyclonal antibodies

  • Non-mammalian-based media composed of recombinant Protein A coupled with Sepharose 4 Fast Flow base matrix by reductive amination
  • Coupling chemistry that minimizes leakage of protein A ligand
  • Multi-point attachment of rProtein A by chemically-stable amide bonds
  • High capacity and good pressure/flow characteristics provide for high throughput

  • Matrix: 4% cross-linked agarose
  • Average particle size: 90μm (d50V)
  • Ligand: recombinant protein A (E. coli)
  • pH Stability working range: 3 to 10
  • pH Stability cleaning-in-place: 2 to 11
  • Binding capacity/mL chromatography medium: >35mg human IgG/mL
  • Pressure/Flow specification: base matrix 150 to 250cm/hr., 100kPa, XK 50/60 column

For therapeutic applications requiring extremely pure eluate fractions of antibodies with non-exposure of antibodies to human or animal derivatives during purification process.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 mL
Stability Stable in aqueous Buffers commonly used in Protein A Chromatography
For Use With (Application) For High-Purity Separation of Monoclonal and Polyclonal Antibodies
Storage Requirements 2°C to 8°C
Binding Capacity >Min. 22 mg Human IgG/mL
Product Type rmp Protein A Sepharose Fast Flow Media
Flow Rate 150 to 250 cm/Hoμr
Particle Size 60 to 165 μm
Height (English) Bed 9.84 in.
Height (Metric) Bed 25 cm
Max. Pressure 100 kPa
Matrix 4% Cross-Linked Agarose
pH Range 3 to 10
Storage Buffer 20% Ethanol
Ligand Type Recombinant Protein A (E. coli)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.