missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Processor Plus Blot Processing Trays

Product Code. 10350845 Shop All Cytiva Products
Change view
Click to view available options
Type:
Blot Processing Mini Tray
Blot Processing Tray Pack
Unit Size:
Each
Pack of 3
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10350845 Blot Processing Mini Tray Pack of 3
10476724 Blot Processing Tray Pack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10350845 Supplier Cytiva Supplier No. 80644442

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

For Blot Processing Tray Pack

  • One tray support
  • Three blot processing mini trays
  • Three blot processing standard trays
  • One blot lid assembly with valves and tubing
  • One blot tray base
  • Four reagent bottles and a rack
  • Two wast bottle caps
  • One spirit level
  • One lid support

For blot processing

Blot Processing Mini Tray

  • Tray volume: 10 to 50mL per chamber
  • Operating temperature: 4° to 40°C

Blot Processing Tray Pack

  • Trays for four mini- (up to 9 x 9.5cm) standard sized blots
  • High-speed peristaltic pump for fast delivery and removal of reagents
  • One 10-port valve for multiple reagent protocols and hazardous waste segregation
  • One rhomboid-shaped tray with smooth and even rocking for rapid fluid mixing and coverage
  • Nine preprogrammed and four user-defined protocols for proteins and nucleic acids
  • Protocol entry and editing, up to 30 steps
  • Removable protocol key for storing and protects critical protocols
TRUSTED_SUSTAINABILITY

Specifications

Description Blot Processing Mini Tray
For Use With (Equipment) Processor Plus™ System for Gel Staining and Blot Processing
Type Blot Processing Mini Tray
Temperature (English) Operating 39.2°F to 104°F
Temperature (Metric) Operating 4°C to 40°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.