missing translation for 'onlineSavingsMsg'
Learn More

Cytiva PhastGel™ 20%, Tris-Acetate, 0.45 mm, Protein Gel

Product Code. 11718468 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10/Pk.
Packungsgröße:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
11718468 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.
Artikelnummer. 11718468 Lieferant Cytiva Lieferanten-Nr. 17062401

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.

Separation Range: 10 to 50 Mr; Separation Type: Denaturing, Native

  • Select from homogeneous and gradient polyacrylamide gels
  • Separate proteins and peptides on denaturing and native
  • Save time and improve reproducibility using precast gels with native or denaturing buffer strips and staining kits, buffer strips, and staining kits
  • Achieve detection sensitivity comparable to that of radioisotopes with ready-to-use silver staining kits
  • Gels are cast on a plastic backing for ease of handling

PhastGel homogenous gels

  • Available in 7.5%, 12.5%, and 20% polyacrylamide with stacking gel, for flexibility in separation, extending the resolution between proteins within a narrow MW range
  • A high density PhastGel (20%) is also available containing PEG to optimise separations below Mr 20000 increasing resolution

PhastGel gradient gels

  • Available in concentration gradients of 4-15%,10-15% and 8-25% with stacking gel, for separation of complex mixtures, giving a good overall view of total composition within a sample
TRUSTED_SUSTAINABILITY

Spezifikation

Boiling Point 125°C
Cassette Material Polyester
Description PhastGel Homogeneous – 20
Gel Percentage 20%
Gel Thickness 0.45 mm
Gel Type Tris-Acetate
Separation of Protein
Separation Range 10 to 50 Mr
Separation Type Denaturing, Native
For Use With (Application) Analytical Chemistry and Research
Material GelBond polyester film
Melting Temperature 82° to 86°C
Molecular Weight (g/mol) 71.08
Quantity 10/Pk.
Stacking Zone 7.5% T/2% C
Storage Requirements 4 - 8°C
Thickness (English) 0.017 in.
Length (Metric) 50 mm
Length (Metric) Gel 5.0 cm
Width (English) 1.9 in.
Width (English) Gel 1.9 in.
Width (Metric) 50 mm
Width (Metric) Gel 5.1 cm
Height (English) 2 in.
Height (Metric) 5.1 cm
Product Type Homogeneous Page
Mehr anzeigen Weniger anzeigen
compliance-icons
Signalwort
  • Danger||
Gefahrenkategorie
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
Gefahrenhinweis
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • EUH208-Contains Acrylamide. May produce an allergic reaction.
Sicherheitshinweis
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Ergänzende Informationen
  • MIXTURE LIST-Contain: Acrylamide
  • UNKNOWN ACUTE-85 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 85% of components with unknown hazards to the aquatic environment.
  • REA17-CMR1-Restricted to professional users
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.