missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Low leakage of protein A with nearly twice total IgG binding capacity of Protein A Sepharose CL-4B
- Used in large-scale FDA-approved processes
- Manufactured without using animal-derived components
- Rigidity and high degree of substitution of Sepharose 4 Fast Flow matrix facilitates speedy process of large volumes dilute cell culture fluid
- High chemical, thermal and mechanical stability
- Withstands high concentrations of hydrogen bond disrupting agents
Specifications
Specifications
| Quantity | 25 mL |
| Stability | No significant change in chromatographic performance after 1 week storage using 8 M Urea, 6 M Gua-HCI, 2% Benzyl Alcohol or 20% Ethanol |
| For Use With (Application) | Purification of Monoclonal and Polyclonal Antibodies at Both Laboratory and Process Scale |
| Storage Requirements | 2°C to 8°C |
| Binding Capacity | >35 mg Human IgG/mL |
| Product Type | nProtein A Sepharose 4 Fast Flow Media |
| Flow Rate | 150 to 250 cm/Hoμr |
| Particle Size | 90 μm |
| Height (English) Bed | 9.84 in. |
| Height (Metric) Bed | 25 cm |
| Show More |
Safety and Handling
Product Identifier
- nProtein A Sepharose 4 Fast Flow, 25 ml
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: ethanol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?