missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- High ligand selectivity and matrix hydrophilicity
- Improved process economics through reduced raw materials costs and/or reduced number of cycles
- Slightly lower pressure/flow properties compared to MabSelect™ SuRe™ and MabSelect
Specifications
Specifications
| Quantity | 200 mL |
| For Use With (Application) | For Develop to Meet The Demands of Increasing Levels of Expression in Monoclonal Antibody Feedstocks |
| Storage Requirements | 2°C to 8°C |
| Binding Capacity | 40 mg Human lgG/mL (at 2.4 min. residence time) |
| Product Type | MabSelect Xtra Media |
| Flow Rate | <300 cm/Hoμr |
| Particle Size | 75 μm |
| Height (English) Bed | 7.87 in. |
| Height (Metric) Bed | 20 cm |
| Max. Pressure | <200 kPa |
| Show More |
Safety and Handling
Product Identifier
- MabSelect Xtra, 200 mL
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?