missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Hot Start Mix RTG

Product Code. 11349941 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
illustra Hot Start Mix RTG
Quantity:
100 Reactions
480 Reactions
8 Reactions
96 Reactions
Volume:
200μL
500μL
Unit Size:
100 reactions
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity Volume unitSize
11349941 illustra Hot Start Mix RTG 8 Reactions 200μL Each
10544625 illustra Hot Start Mix RTG 96 Reactions 200μL Each
10126624 illustra Hot Start Mix RTG 100 Reactions 500μL 100 reactions
10625985 illustra Hot Start Mix RTG 480 Reactions 200μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11349941 Supplier Cytiva Supplier No. 28904091

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Premixed, predispensed, ambient temperature-stable formulation providing robust and reliable performance for demanding PCR applications in which high specificity and high sensitivity are essential to success

  • Primer sequestration effectively blocks greater than 99% of polymerase activity during setup effectively minimizing any nonspecific priming or primer-dimer formation
  • Recombinant activator protein avoids the risk of contamination from a mammalian source associated with hot start antibodies
  • Quick hot start activation preserves sample integrity by avoiding extensive precycling heat treatment
  • The Ready-To-Go manufacturing process locks in performance for excellent reproducibility with less risk of pipetting errors and contamination
  • Long-term ambient-temperature stability simplifies shipping and removes any need for freezer storage

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1.75 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer. Ambient temperature in pouch with desiccant
Format Tube
Product Type illustra Hot Start Mix RTG
Quantity 8 Reactions
For Use With (Application) Standard PCR
No. of Reactions 8
Volume 200μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.