missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Dithiothreitol (DTT)

Include reducing agents in the sample solution to break any disulfide bonds present and to maintain all proteins in their fully reduced state. Cytiva Life Sciences™ Dithiothreitol (DTT) is one of the most commonly used reductants.

Specifications

For Use With (Application) IEF, SDS-PAGE
pH 0 to 5.1
View More Specs

Description

Description

  • Use for IEF, SDS-PAGE
  • White
  • Solid
  • Easily soluble in cold water and hot water
  • Threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT
Specifications

Specifications

IEF, SDS-PAGE
0 to 5.1
Videos
Safety and Handling

Safety and Handling

Signal Word
  • ||Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications