missing translation for 'onlineSavingsMsg'
Learn More

Cytiva cAMP Direct BioTrak™ EIA

Use for direct measurement of cAMP in cell lysates without any need for removal of extraction reagents. Cytiva Life Sciences™ cAMP Direct BioTrak™ EIA represents a convenient, high-sensitivity, peroxidase-based technology that enables the amount of protein present in a sample to be measured in a single day.

£545.00 - £2432.00

Specifications

Product Type cAMP biotrak Enzyme immunoassay (EIA) system
For Use With (Application) Used for research purposes, eliminates lengthy sample extraction procedures and allow direct cAMP measurement
Storage Requirements 2°C to 8°C
View More Specs

Products 2
Product Code Brand Quantity Assay Sensitivity Price Quantity & Availability  
Product Code Brand Quantity Assay Sensitivity Price Quantity & Availability  
10451835
View Documents Promotion Details
Cytiva
RPN2251
96 Determinations 38.4pg/mL (non-acetylation) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11798358
View Documents Promotion Details
Cytiva
RPN2255
5 x 96 Determinations 38.4pg/mL (non-acetylation), 14pg/mL (acetylation) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • Flexible method-choice of assay protocols
  • Rapid assay protocol
  • Non-radioactive
  • Precise and accurate measurement
  • Ready-to-use substrate
  • Color coded reagents
  • Five-hour protocol

  • Kits available with or without acetylation reagents
  • Bulk kit provided without lysis buffers
  • Dual range: 25–6400 fmol/well [non-acetylation], 2–128 fmol/well [acetylation]
  • Sensitivity: 38.4pg/mL [non-acetylation] and 14pg/mL [acetylation]

Specifications

Specifications

cAMP biotrak Enzyme immunoassay (EIA) system
Used for research purposes, eliminates lengthy sample extraction procedures and allow direct cAMP measurement
2°C to 8°C
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P270-Do not eat, drink or smoke when using this product.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P264-Wash hands thoroughly after handling.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P405-Store locked up.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : calcium chloride
  • UNKNOWN ACUTE-100 (kit) per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 100 (kit)% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Product Certifications