missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Rapid-hyb Buffer

Product Code. 11585055 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11585055 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11585055 Supplier Cytiva Supplier No. RPN1636

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Pre-mixed, rate-enhancing hybridization buffer. Amersham™ Rapid-hyb Buffer reduces hybridization time for detecting single-copy mammalian genes to just two hours.

  • For use with radiolabeled DNA, RNA, or oligonucleotide probes
  • Pre-mixed solution improves reproducibility between experiments
  • Suited for Southern and Northern blotting applications
  • Available in two different quantities, sufficient for 1000cm2 membrane and 4000cm2 membrane

Developed for use with nylon membranes; not suitable for use with PVDF or nitrocellulose membranes.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at ambient temperature. Under these conditions the product is stable for at least 3 months
For Use With (Application) Radiolabeled DNA, RNA or Oligonucleotide probes
Product Type Rapid-hyb Buffer sufficient for 4000 cm2 membrane
Quantity 500 mL
compliance-icons
Product Identifier
  • Rapid-hyb Buffer sufficient for 4,000 cm2 membrane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H319-Causes serious eye irritation.
Supplemental information
  • MIXTURE LIST-Contain: 2-hydroxy-1,2,3-Propanetricarboxylic acid hydrate
  • UNKNOWN ACUTE-19 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 6% of components with unknown hazards to the aquatic environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.