missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Amersham Hypercassette autoradiography cassettes are constructed from strong aluminum and designed for the toughest demands of life science research. Deep cassettes are 1.5cm in depth, allowing them to accommodate thicker samples. A choice of Hypercassette colors enables color coding of work.
- Provide maximum contact between film and sample
- Nonporous pad gives a wipe clean surface, ensuring minimum contamination
Specifications
Specifications
| Product Type | Hypercassette, Neutral (Standard) |
| Description | Amersham hypercssette autoradiography cassettes are designed to survive the toughest demands of research in the life sciences. |
| Color | Neutral (Standard) |
| For Use With (Application) | Light-tight exposures at temperatures below -70°C |
| Dimensions (LxW) | 24 x 30 cm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?