missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDPD5 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41268MFV610
This item is not returnable.
View return policy
Description
GDPD5 Polyclonal antibody specifically detects GDPD5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| GDPD5 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| EC 3.1, GDE2PP1665Glycerophosphodiester phosphodiesterase 2, glycerophosphodiester phosphodiesterase domain containing 5, glycerophosphodiester phosphodiesterase domain-containing protein 5 | |
| Antibody was raised against an 18 amino acid peptide near the amino terminus of human GDPD5. The immunogen is located within the first 50 amino acids of GDPD5. Amino Acid Squence: RWKRYQRSHDDTTPWER | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 81544 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction