missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDPD5 Antibody (506020) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10524JF635
This item is not returnable.
View return policy
Description
GDPD5 Monoclonal antibody specifically detects GDPD5 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| GDPD5 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| EC 3.1, GDE2PP1665Glycerophosphodiester phosphodiesterase 2, glycerophosphodiester phosphodiesterase domain containing 5, glycerophosphodiester phosphodiesterase domain-containing protein 5 | |
| Human embryonic kidney cell, HEK293-derived transfected with GDPD5, Accession # Q8WTR4 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 506020 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 81544 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction