missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GDPD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113341
This item is not returnable.
View return policy
Description
GDPD2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Has glycerophosphoinositol inositolphosphodiesterase activity and specifically hydrolyzes glycerophosphoinositol, with no activity for other substrates such as glycerophosphoinositol 4-phosphate, glycerophosphocholine, glycerophosphoethanolamine, and glycerophosphoserine. Accelerates the program of osteoblast differentiation and growth. May play a role in remodeling of the actin cytoskeleton.
Specifications
| GDPD2 | |
| Polyclonal | |
| Unconjugated | |
| GDPD2 | |
| 9130017L10Rik; EC 3.1.4.43; FLJ20207; Gde3; GDPD2; Glycerophosphodiester phosphodiesterase 3; glycerophosphodiester phosphodiesterase domain containing 2; glycerophosphodiester phosphodiesterase domain-containing protein 2; Glycerophosphoinositol inositolphosphodiesterase GDPD2; OBDPF; Osteoblast differentiation promoting factor; RGD1560780; UNQ1935/PRO4418 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54857 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 4.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9HCC8 | |
| GDPD2 | |
| Recombinant Human Glycerophosphoinositol inositolphosphodiesterase GDPD2 protein (210-490AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction