missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gdpd1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570901
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%.
Hydrolyzes lysoglycerophospholipids to produce lysophosphatidic acid (LPA) and the corresponding amines. Shows a preference for 1-O-alkyl-sn-glycero-3-phosphocholine (lyso-PAF), lysophosphatidylethanolamine (lyso-PE) and lysophosphatidylcholine (lyso-PC). May be involved in bioactive N-acylethanolamine biosynthesis. Does not display glycerophosphodiester phosphodiesterase activity, since it cannot hydrolyze either glycerophosphoinositol or glycerophosphocholine.
Specifications
| Gdpd1 | |
| Polyclonal | |
| Unconjugated | |
| Gdpd1 | |
| 2610020H15Rik; GDE4; GDPD1; Glycerophosphodiester phosphodiesterase 4; glycerophosphodiester phosphodiesterase domain containing 1; glycerophosphodiester phosphodiesterase domain-containing protein 1; Lysophospholipase D GDPD1; RGD1311813; Unknown (protein for MGC:134023) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 303407 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q0VGK4 | |
| Gdpd1 | |
| synthetic peptide directed towards the N-terminal region of rat Gdpd1. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction