missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GDNF Recombinant Rabbit Monoclonal Antibody (JA93-10)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA541229

Product Code. 17451383

  • £453.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GDNF Recombinant Monoclonal Antibody for Western Blot, ICC/IF

GDNF is a glycosylated, disulfide-bonded homodimer molecule. It was first discovered as a potent survival factor for midbrain dopaminergic neurons and was then shown to rescue these neurons in animal models of Parkinson's disease. GDNF is about 100 times more efficient survival factor for spinal motor neurons than the neurotrophins. GDNF is a neurotrophic factor that enhances survival and morphological differentiation of dopaminergic neurons and increases their high-affinity dopamine uptake.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GDNF
Recombinant Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
P39905, P48540, Q07731
GDNF
Synthetic peptide within human GDNF aa 51-100/211.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Western Blot, Immunocytochemistry
JA93-10
Unconjugated
GDNF
AI385739; Astrocyte-derived trophic factor; ATF; ATF1; ATF2; Gdnf; glial cell derived neurotrophic factor; glial cell line derived neurotrophic factor; Glial cell line derived neutrophic factor; glial cell line-derived neurotrophic factor; gndf; HFB1-GDNF; hGDNF; H-GDNF; HSCR3; mGDNF; neurotrophic factor
Rabbit
Protein A
RUO
14573, 25453, 2668
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktförslag

Produktförslag

Filmer
SDS
Dokument

Dokument

Produktspecifikationer
Kampanjer

Kampanjer

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.