Learn More
PeproTech GDNF Polyclonal Antibody, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
£177.00 - £1468.00
Specifications
| Antigen | GDNF |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17853353
![]() |
PeproTech
500-P81-50UG |
50 μg |
£177.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17863353
![]() |
PeproTech
500-P81-100UG |
100 μg |
£228.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17843723
![]() |
PeproTech
500-P81-1MG |
1 mg |
£1468.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
AA Sequence of recombinant protein: MSPDKQMAVL PRRERNRQAA AANPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCDAAETTY DKILKNLSRN RRLVSDKVGQ ACCRPIAFDD DLSFLDDNLV YHILRKHSAK RCGCI. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human GDNF. Anti-Human GDNF-specific antibody was purified by affinity chromatography employing an immobilized hHuman GDNF matrix. Sandwich ELISA: To detect Human GDNF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human GDNF (500-P81Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human GDNF. Western Blot: To detect Human GDNF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human GDNF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
GDNF is a glycosylated, disulfide-bonded homodimer molecule. It was first discovered as a potent survival factor for midbrain dopaminergic neurons and was then shown to rescue these neurons in animal models of Parkinson's disease. GDNF is about 100 times more efficient survival factor for spinal motor neurons than the neurotrophins. GDNF is a neurotrophic factor that enhances survival and morphological differentiation of dopaminergic neurons and increases their high-affinity dopamine uptake.Specifica
| GDNF | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P39905 | |
| 2668 | |
| E.coli-derived Recombinant Human GDNF. | |
| Antigen affinity chromatography | |
| GDNF |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| AI385739; Astrocyte-derived trophic factor; ATF; ATF1; ATF2; Gdnf; glial cell derived neurotrophic factor; glial cell line derived neurotrophic factor; Glial cell line derived neutrophic factor; glial cell line-derived neurotrophic factor; gndf; HFB1-GDNF; hGDNF; H-GDNF; HSCR3; mGDNF; neurotrophic factor | |
| GDNF | |
| Primary | |
| -20°C |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.