missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDI2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55096-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to GDI2 and it has no cross reaction to GDI1.
Regulates the GDP/GTP exchange reaction of most Rab proteins by inhibiting the dissociation of GDP from them, and the subsequent binding of GTP to them.Specifications
| GDI2 | |
| Polyclonal | |
| Unconjugated | |
| GDI2 | |
| GDI 2, GDI2, GDP dissociation inhibitor 2, Rab GDI beta, RABGDIB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2665 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| 0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P50395 | |
| GDI2 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction