missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10116-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody can bind both GDIs for the close sequences.
Regulates the GDP/GTP exchange reaction of most Rab proteins by inhibiting the dissociation of GDP from them, and the subsequent binding of GTP to them.Specifications
| GDI2 | |
| Polyclonal | |
| Unconjugated | |
| GDI2 | |
| GDI 2, GDI2, GDP dissociation inhibitor 2, Rab GDI beta, RABGDIB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14569, 2665, 29662 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P50395, P50399, Q61598 | |
| Gdi2 | |
| GDI2 Fusion Protein Ag0170 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction