missing translation for 'onlineSavingsMsg'
Learn More

GDI1 Antibody, HRP, Novus Biologicals™

Product Code. 18354755 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354755 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354755 Supplier Novus Biologicals Supplier No. NBP299205H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GDI1 Polyclonal specifically detects GDI1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDI1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias FLJ41411, GDI-1, GDIL, GDP dissociation inhibitor 1, Guanosine diphosphate dissociation inhibitor 1, mental retardation, X-linked 41, mental retardation, X-linked 48, MRX41, oligophrenin-2, OPHN2MRX48, Protein XAP-4, Rab GDI alpha, rab GDP dissociation inhibitor alpha, rab GDP-dissociation inhibitor, alpha, RABGDIARABGD1A, XAP4, XAP-4
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human GDI1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2664
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.