missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GDF7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106668
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GDF7.
This gene encodes a member of the bone morphogenetic protein (BMP) family. BMPs belong to the transforming growth factor-beta superfamily of secreted signalling molecules that regulate diverse processes in growth, repair and embryonic development. In mouse, this gene functions as an inductive signal from the roof plate required for the specification of neuronal identity in the dorsal spinal cord.
Specifications
| GDF7 | |
| Polyclonal | |
| Unconjugated | |
| GDF7 | |
| BMP12; Gdf7; Gdf-7; growth differentiation factor 7; growth/differentiation factor 7 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 151449 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q7Z4P5 | |
| GDF7 | |
| A synthesized peptide derived from human GDF7(Accession Q7Z4P5), corresponding to amino acid residues L194-P244. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction