missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GDF5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529775
This item is not returnable.
View return policy
Description
Recommended positive controls: IMR32. Predicted reactivity: Human (99%), Mouse (96%), Pig (98%), Rabbit (98%), Chicken (83%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GDF-5 is expressed in long bones during embryonic development and postnatally in articular cartilage. Mutations in the GDF-5 gene have been implicated in Hunter-Thompson type dwarfism and in Grebe Syndrome, which is characterized by short stature, extra digits, and short and deformed extremities. The mature and functional form of GDF-5 is a homodimer of two 120 amino-acid polypeptide chain (monomers) linked by a single disulfide bond. Each GDF-5 monomer is expressed as the C-terminal part of a precursor polypeptide, which also contains a 27 amino acid signal peptide and a 354 amino acid propeptide. This precursor undergoes intracellular dimerization, and upon secretion it is processed by a furin-type protease.
Specifications
| GDF5 | |
| Polyclonal | |
| Unconjugated | |
| GDF5 | |
| BDA1C; Bmp14; BMP-14; Bone morphogenetic protein 14; bp; brachypodism; brp; Cartilage-derived morphogenetic protein 1; cartilage-derived morphogenetic protein-1; CDMP1; Cdmp-1; GDF5; GDF-5; growth differentiation factor 5; growth/differentiation factor 5; H-GDF-5; LAP4; LAP-4; Lipopolysaccharide-associated protein 4; LPS-associated protein 4; OS5; radotermin; SYM1B; SYNS2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8200 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Functional Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.65 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P43026 | |
| GDF5 | |
| Recombinant protein encompassing a sequence within the center region of human GDF5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction