missing translation for 'onlineSavingsMsg'
Learn More

PeproTech GDF3 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

£177.00 - £2745.00

Specifications

Antigen GDF3
Concentration 0.1-1.0 mg/mL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17895813
View Documents
PeproTech
500-P235BT-25UG
25 μg
£177.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17855793
View Documents
PeproTech
500-P235BT-50UG
50 μg
£228.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17845793
View Documents
Invitrogen
500-P235BT-1MG
2 x 500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
30054693
View Documents
PeproTech
500-P235BT-500UG
500 μg
£1466.00
500µg
Se alternative produkter
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Beskrivelse

Beskrivelse

AA Sequence of recombinant protein: AAIPVPKLSC KNLCHRHQLF INFRDLGWHK WIIAPKGFMA NYCHGECPFS LTISLNSSNY AFMQALMHAV DPEIPQAVCI PTKLSPISML YQDNNDNVIL RHYEDMVVDE CGCG. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human GDF-3. Anti-Human GDF-3-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hGDF-3 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human GDF-3 (500-P235) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hGDF-3. Western Blot: To detect hGDF-3 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hGDF-3 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P235BT-1MG will be provided as 2 x 500 μg

GDF3 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. The function of this protein is unknown, but expression studies suggest it may be involved in regulation of the adult lymphatic and erythroid systems and embryonic development.
Tekniske data

Tekniske data

GDF3
ELISA, Western Blot
Biotin
Rabbit
Human
Q9NR23
9573
E.coli-derived Recombinant Human GDF-3.
Antigen affinity chromatography
GDF3
0.1-1.0 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative
C78318; ecat9; Gdf3; Gdf-3; growth differentiation factor 3; growth/differentiation factor 3; KFS3; MCOP7; MCOPCB6; RGD1564178; UNQ222/PRO248; VG-1-related protein 2; Vgr2; Vgr-2
GDF3
Primary
-20°C
Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.