missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-9 Antibody (917319), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8266PECY55
This item is not returnable.
View return policy
Description
GDF-9 Monoclonal antibody specifically detects GDF-9 in Human samples. It is validated for Flow Cytometry
Specifications
| GDF-9 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 917319 | |
| Flow Cytometry | |
| GDF9, GDF-9, growth differentiation factor 9, growth/differentiation factor 9 | |
| Human embryonic kidney cell line derived recombinant human GDF-9, Met1-Arg454, Accession # O60383 | |
| 0.1 mL | |
| Cytokine Research | |
| 2661 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction