missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-9 Antibody (72B), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-61933H
This item is not returnable.
View return policy
Description
GDF-9 Monoclonal specifically detects GDF-9 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| GDF-9 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| GDF9 | |
| N-terminal of GDF-9 corresponding to sequence KKPLGPASFNLSEYFC | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| 72B | |
| Western Blot, ELISA | |
| GDF9, GDF-9, growth differentiation factor 9, growth/differentiation factor 9 | |
| Rat | |
| Protein G purified | |
| Cytokine Research | |
| 2661 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction