missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-9 Antibody (53/1) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£159.00 - £378.00
Specifications
| Antigen | GDF-9 |
|---|---|
| Clone | 53/1 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18200833
|
Novus Biologicals
NBP2-61934 |
0.1 mg |
£378.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18693318
|
Novus Biologicals
NBP2-61934-0.025mg |
0.025 mg |
£159.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GDF-9 Monoclonal specifically detects GDF-9 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
| GDF-9 | |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| PBS with 0.02% Sodium Azide | |
| 2661 | |
| Tuberculin coupled peptide with sequence VPAKYSPLSVLTIEPDGSIAYKEYEDMIATKC that recognizes an epitope with the EPDG sequence near the C-terminal region of human GDF-9 | |
| Primary |
| 53/1 | |
| Monoclonal | |
| Purified | |
| Cytokine Research | |
| GDF9, GDF-9, growth differentiation factor 9, growth/differentiation factor 9 | |
| GDF9 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title