missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ GDF-8/Myostatin Quantikine ELISA Kit
Click to view available options
Sufficient For:
1 x 96-well Plate
Unit Size:
1 set
Description
The GDF-8/Myostatin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies multi-species GDF-8/Myostatin in cell culture supernates (100 ul), tissue homogenates (20 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul).
Specifications
Specifications
| Assay Range | 31.30 - 2000pg/mL |
| Assay Sensitivity | 5.32pg/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Homogenates |
| Specificity | Natural and Recombinant Human, Mouse, Rat, Canine, Porcine, and equine GDF-8. |
| Sufficient For | 1 x 96-well Plate |
| For Use With (Application) | ELISA |
| Sample Volume | Cell Culture Supernates (100uL),Tissue Homogenates (20uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Synonym | GDF8, GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, MSLHP, MSTN, Myostatin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction