missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-8/Myostatin Mouse anti-Human, Clone: GDF9/4261, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07706-20UG
This item is not returnable.
View return policy
Description
GDF-8/Myostatin Monoclonal specifically detects GDF-8/Myostatin in Human samples. It is validated for Western Blot, ELISA.
Specifications
| GDF-8/Myostatin | |
| Monoclonal | |
| 0.2 mg/ml | |
| Western Blot 1-2 ug/ml, ELISA | |
| GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, Myostatin | |
| Recognizes the 113 amino acid carboxy-terminal fragment of GDF-8/Myostatin protein | |
| 20 μg | |
| Cytokine Research | |
| 2660 | |
| Store at 4°C. | |
| IgG |
| Western Blot, ELISA | |
| GDF9/4261 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction