missing translation for 'onlineSavingsMsg'
Learn More

GDF-6/BMP-13 Antibody [PerCP], Novus Biologicals™

Product Code. 30732548 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732548 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732548 Supplier Novus Biologicals Supplier No. AF855C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

GDF-6/BMP-13 Polyclonal antibody specifically detects GDF-6/BMP-13 in Mouse samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDF-6/BMP-13
Applications Immunohistochemistry
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias BMP13, CDMP2, GDF16, GDF-6, growth differentiation factor 6, Growth/differentiation factor 16, growth/differentiation factor 6, KFS, KFS1, KFSL, MCOP4, MCOPCB6, MGC158100, MGC158101, SCDO4, SGM1
Host Species Sheep
Immunogen E. coli-derived recombinant mouse GDF-6/BMP-13, Thr335-Arg454, Accession # P43028
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Vision
Primary or Secondary Primary
Gene ID (Entrez) 392255
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.