missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-6/BMP-13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-91934-25ul
This item is not returnable.
View return policy
Description
GDF-6/BMP-13 Polyclonal specifically detects GDF-6/BMP-13 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| GDF-6/BMP-13 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| BMP13, CDMP2, GDF16, GDF-6, growth differentiation factor 6, Growth/differentiation factor 16, growth/differentiation factor 6, KFS, KFS1, KFSL, MCOP4, MCOPCB6, MGC158100, MGC158101, SCDO4, SGM1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| GDF6 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:IYRTYSIAEKLGINASFFQSSKSANTITSFVDRGLDDLSHTPLRRQKYLFDVSMLSDKEELVGAELRLFRQAPSAPWGPPAGPLHVQLF | |
| 25 μL | |
| Vision | |
| 392255 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction