missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-6/BMP-13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-87494
This item is not returnable.
View return policy
Description
GDF-6/BMP-13 Polyclonal specifically detects GDF-6/BMP-13 in Human samples. It is validated for Western Blot.
Specifications
| GDF-6/BMP-13 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| BMP13, CDMP2, GDF16, GDF-6, growth differentiation factor 6, Growth/differentiation factor 16, growth/differentiation factor 6, KFS, KFS1, KFSL, MCOP4, MCOPCB6, MGC158100, MGC158101, SCDO4, SGM1 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of human GDF-6/BMP-13 | |
| 100 μL | |
| Biologically Active Proteins, Vision | |
| 392255 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction