missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-6/BMP-13 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76364MFV610
This item is not returnable.
View return policy
Description
GDF-6/BMP-13 Polyclonal antibody specifically detects GDF-6/BMP-13 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| GDF-6/BMP-13 | |
| Polyclonal | |
| Western Blot, ELISA | |
| BMP13, CDMP2, GDF16, GDF-6, growth differentiation factor 6, Growth/differentiation factor 16, growth/differentiation factor 6, KFS, KFS1, KFSL, MCOP4, MCOPCB6, MGC158100, MGC158101, SCDO4, SGM1 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of the human GDF6. The immunogen is located within amino acids 320 - 370 of GDF6. Amino Acid Squence: AFASRHGKRHGKKSRLR | |
| 0.1 mL | |
| Biologically Active Proteins, Vision | |
| 392255 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction