missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-5/BMP-14 Antibody (RM0242-13J15), mFluor Violet 450 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12333MFV450
This item is not returnable.
View return policy
Description
GDF-5/BMP-14 Monoclonal antibody specifically detects GDF-5/BMP-14 in Mouse samples. It is validated for Western Blot, Neutralization
Specifications
| GDF-5/BMP-14 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Neutralization | |
| RM0242-13J15 | |
| Western Blot, Block/Neutralize | |
| BMP14, Cartilage-derived morphogenetic protein 1, cartilage-derived morphogenetic protein-1, CDMP-1, CDMP1LAP4, GDF-5, growth differentiation factor 5, growth/differentiation factor 5, OS5, Radotermin, SYNS2 | |
| Mouse recombinant GDF-5/BMP-14 | |
| 0.1 mL | |
| Cytokine Research | |
| 8200 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction